Learn More
Incell Corporation ZAPR RBC Lysing Buffer, 100 mL, sterile, chemically defined, xeno-free, endotoxin <0.1 EU/mL, pH 7.2-7.6, shelf life 24 months
SDP
Catalog No. NC1660620
Encompass
ZAPR RBC Lysing Buffer, 100 mL, sterile, chemically defined, xeno-free, endotoxin <0.1 EU/mL, pH 7.2-7.6, shelf life 24 months
Catalog No. NC1660620 Supplier Incell Corporation Supplier No. ZAPR100
ZAPR RBC Lysing Buffer, 100 mL, sterile, chemically defined, xeno-free, endotoxin <0.1 EU/mL, pH 7.2-7.6, shelf life 24 months