Learn More
Medchemexpress LLC Zaprinast | 37762-06-4 | 99.88% | 50 MG
SDP
Catalog No. 5000412762
Encompass_Preferred
Zaprinast (M&B 22948) is a selective inhibitor of cGMP-selective Phosphodiesterase (PDE5) that significantly increases cGMP levels in myocytes. It is also a G protein-coupled receptor 35 (GPR35) agonist, activating rat GPR35 strongly and human GPR35 moderately. Zaprinast reduces vessel remodeling through antiproliferative and proapoptotic effects.
  • Selective inhibitor of cGMP-selective Phosphodiesterase (PDE5).
  • Increases cGMP levels in myocytes.
  • G protein-coupled receptor 35 (GPR35) agonist.
  • Reduces vessel remodeling through antiproliferative and proapoptotic effects.
  • Enhances spatial memory and diminishes exploratory activity in mice.
Catalog No. 50-004-12762 Supplier Medchemexpress LLC Supplier No. HYB181650MG
Zaprinast (M&B 22948) is a selective inhibitor of cGMP-selective Phosphodiesterase (PDE5) that significantly increases cGMP levels in myocytes. It is also a G protein-coupled receptor 35 (GPR35) agonist, activating rat GPR35 strongly and human GPR35 moderately. Zaprinast reduces vessel remodeling through antiproliferative and proapoptotic effects.
  • Selective inhibitor of cGMP-selective Phosphodiesterase (PDE5).
  • Increases cGMP levels in myocytes.
  • G protein-coupled receptor 35 (GPR35) agonist.
  • Reduces vessel remodeling through antiproliferative and proapoptotic effects.
  • Enhances spatial memory and diminishes exploratory activity in mice.