Learn More
Abcam Zaprinast, cGMP phosphodiesterase inhibitor, 10MG
Catalog No. 502707105
Encompass_Preferred
MW 271.28 g/mol, Purity >98%. Specific cGMP phosphodiesterase inhibitor, selective for PDE6, 5, 11 and 9 (IC₅₀ values are 0.15 (PDE6), 0.76 (PDE5), 12 (PDE11) and 29 μM (PDE9)). Reputed GPR35 agonist (EC₅₀ values are 16 and 840 nM at rat and human GPR35 receptors, respectively). Important mediator for a variety of cellular processes including vasodilation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7105 Supplier Abcam Supplier No. AB12094010MG
MW 271.28 g/mol, Purity >98%. Specific cGMP phosphodiesterase inhibitor, selective for PDE6, 5, 11 and 9 (IC₅₀ values are 0.15 (PDE6), 0.76 (PDE5), 12 (PDE11) and 29 μM (PDE9)). Reputed GPR35 agonist (EC₅₀ values are 16 and 840 nM at rat and human GPR35 receptors, respectively). Important mediator for a variety of cellular processes including vasodilation.
The product is subject to the following: Abcam Restricted Use Statement