Learn More
Medchemexpress LLC Zasocitinib | 2272904-53-5 | 99.2% | 460.49 | 1 ML
SDP
Catalog No. 5000361085
Encompass_Preferred
Zasocitinib (NDI-034858) is a TYK2 inhibitor, specifically targeting the TYK2 JH2 domain. It exhibits a binding constant (Kd) of less than 200 pM and is intended for research use only.
  • Targets TYK2 JH2 domain.
  • Binding constant (Kd) of <200 pM.
  • Inhibits TYK2 and JAK at 10 μM.
  • Inhibits phosphorylation induction of heterodimer complex TYK2/JAK2-mediated IL-12 effect in human peripheral blood mononuclear cells with an IC50 value of <0.1 μM.
Catalog No. 50-003-61085 Supplier Medchemexpress LLC Supplier No. HY15009610MM/1ML
Zasocitinib (NDI-034858) is a TYK2 inhibitor, specifically targeting the TYK2 JH2 domain. It exhibits a binding constant (Kd) of less than 200 pM and is intended for research use only.
  • Targets TYK2 JH2 domain.
  • Binding constant (Kd) of <200 pM.
  • Inhibits TYK2 and JAK at 10 μM.
  • Inhibits phosphorylation induction of heterodimer complex TYK2/JAK2-mediated IL-12 effect in human peripheral blood mononuclear cells with an IC50 value of <0.1 μM.