Learn More
Medchemexpress LLC Zasocitinib | 2272904-53-5 | 99.24% | 460.49 | 5 MG
SDP
Catalog No. 5000361935
Encompass_Preferred
Zasocitinib (NDI-034858) is a potent TYK2 inhibitor, specifically targeting the TYK2 JH2 domain with a high binding constant (Kd) of <200 pM. It also exhibits inhibitory effects on Tyk2 and JAK by targeting their JH1 domains. This compound is intended for research use only.
  • Inhibits phosphorylation induction of heterodimer complex Tyk2/JAK2-mediated IL-12 effect
  • IC50 value of <0.1 μM in human peripheral blood mononuclear cells
  • Off-white to light yellow solid appearance
Catalog No. 50-003-61935 Supplier Medchemexpress LLC Supplier No. HY1500965MG
Zasocitinib (NDI-034858) is a potent TYK2 inhibitor, specifically targeting the TYK2 JH2 domain with a high binding constant (Kd) of <200 pM. It also exhibits inhibitory effects on Tyk2 and JAK by targeting their JH1 domains. This compound is intended for research use only.
  • Inhibits phosphorylation induction of heterodimer complex Tyk2/JAK2-mediated IL-12 effect
  • IC50 value of <0.1 μM in human peripheral blood mononuclear cells
  • Off-white to light yellow solid appearance