Learn More
Medchemexpress LLC ZASOCITINIB NDI-034858 1MG
SDP
Supplier:  Medchemexpress LLC HY1500961MG
Encompass_Preferred
Zasocitinib (NDI-034858) is a TYK2 inhibitor, target TYK2 JH2 domain with binding constant Kd of <200 pM.
Catalog No. NC2268020