Learn More
Medchemexpress LLC Zastaprazan | 2133852-18-1 | 100.0% | 362.47 | 50 MG
SDP
Catalog No. 5000349584
Encompass_Preferred
Zastaprazan (JP-1366) is a proton pump inhibitor and a potent potassium-competitive acid blocker. It is suitable for use in the research of gastrointestinal inflammatory diseases or gastric acid-related conditions such as gastroesophageal reflux disease. This product is for research use only.
  • Proton pump inhibitor
  • Potent potassium-competitive acid blocker
  • Used in gastrointestinal inflammatory disease research
  • Used in gastric acid-related disease research
  • For research use only
Catalog No. 50-003-49584 Supplier Medchemexpress LLC Supplier No. HY13955750MG
Zastaprazan (JP-1366) is a proton pump inhibitor and a potent potassium-competitive acid blocker. It is suitable for use in the research of gastrointestinal inflammatory diseases or gastric acid-related conditions such as gastroesophageal reflux disease. This product is for research use only.
  • Proton pump inhibitor
  • Potent potassium-competitive acid blocker
  • Used in gastrointestinal inflammatory disease research
  • Used in gastric acid-related disease research
  • For research use only