Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zastaprazan | 2133852-18-1 | 100.0% | C22H26N4O | 10MG
SDP

Catalog No. 5000261548
Encompass_Preferred

Zastaprazan is a potassium-competitive proton pump inhibitor research compound used to investigate gastrointestinal inflammatory conditions and gastric acid-related diseases such as gastroesophageal reflux disease. It is supplied as a high-purity small molecule with characterized molecular properties.

  • Potent potassium-competitive acid blocker.
  • Used to research gastrointestinal inflammatory and gastric acid-related diseases.
  • High purity (99.96%).
  • Molecular formula C22H26N4O; molecular weight 362.47.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-61548 Supplier Medchemexpress LLC Supplier No. HY13955710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zastaprazan is a potassium-competitive proton pump inhibitor research compound used to investigate gastrointestinal inflammatory conditions and gastric acid-related diseases such as gastroesophageal reflux disease. It is supplied as a high-purity small molecule with characterized molecular properties.

  • Potent potassium-competitive acid blocker.
  • Used to research gastrointestinal inflammatory and gastric acid-related diseases.
  • High purity (99.96%).
  • Molecular formula C22H26N4O; molecular weight 362.47.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.