Learn More
Medchemexpress LLC Zastaprazan | 2133852-18-1 | MFCD34567796 | 100.0% | 362.47 g·mol-1 | C22H26N4O | 5 MG
SDP
Catalog No. 5000221329
Encompass_Preferred
Zastaprazan (JP-1366) is a research-stage proton pump inhibitor that functions as a potassium-competitive acid blocker. It is used in preclinical research on gastric acid-related disorders and gastrointestinal inflammatory disease models, and is supplied as a solid powder for experimental use.
  • Potent potassium-competitive acid blocker mechanism of action.
  • Suitable for in vitro and in vivo research on gastric acid secretion and related disorders.
  • High purity suitable for research applications.
  • Supplied as a powder and available in mg-scale quantities for laboratory use.
  • Stable under frozen storage; follow supplier storage recommendations.
  • Chemical formula C22H26N4O; molecular weight 362.47 g·mol-1.
Catalog No. 50-002-21329 Supplier Medchemexpress LLC Supplier No. HY1395575MG
Zastaprazan (JP-1366) is a research-stage proton pump inhibitor that functions as a potassium-competitive acid blocker. It is used in preclinical research on gastric acid-related disorders and gastrointestinal inflammatory disease models, and is supplied as a solid powder for experimental use.
  • Potent potassium-competitive acid blocker mechanism of action.
  • Suitable for in vitro and in vivo research on gastric acid secretion and related disorders.
  • High purity suitable for research applications.
  • Supplied as a powder and available in mg-scale quantities for laboratory use.
  • Stable under frozen storage; follow supplier storage recommendations.
  • Chemical formula C22H26N4O; molecular weight 362.47 g·mol-1.