Learn More
Medchemexpress LLC Zastaprazan (JP-1366) | 2133852-18-1 | 100.0% | 362.47 | 100 MG
SDP
Catalog No. 5000347970
Encompass_Preferred
Zastaprazan (JP-1366) is a proton pump inhibitor and a potent potassium-competitive acid blocker. It can be used in the research of gastrointestinal inflammatory diseases or gastric acid-related diseases such as gastroesophageal reflux disease.
  • IC50 & Target: proton pump
  • In vitro: Is a proton pump inhibitor
Catalog No. 50-003-47970 Supplier Medchemexpress LLC Supplier No. HY139557100MG
Zastaprazan (JP-1366) is a proton pump inhibitor and a potent potassium-competitive acid blocker. It can be used in the research of gastrointestinal inflammatory diseases or gastric acid-related diseases such as gastroesophageal reflux disease.
  • IC50 & Target: proton pump
  • In vitro: Is a proton pump inhibitor