Learn More
Medchemexpress LLC Zatebradine | 91940-87-3 | 99.9% | C26H37ClN2O5 | 10MG
SDP
Catalog No. 5000261860
Encompass_Preferred
Zatebradine hydrochloride is a research-grade small molecule used to inhibit hyperpolarization-activated cyclic nucleotide-gated (HCN) channels in electrophysiology and pharmacology studies. It blocks HCN1-HCN4-mediated currents with low-micromolar potency and is supplied in solid and solution formats for in vitro experiments.
  • Potent HCN channel antagonist with low-micromolar IC50 values.
  • Blocks HCN1-HCN4-mediated slow inward current.
  • Available as solid material and as a 10 mM solution in DMSO for convenience.
  • High reported purity suitable for research applications.
  • Applicable to electrophysiology, cardiac research, and ion channel pharmacology studies.
Catalog No. 50-002-61860 Supplier Medchemexpress LLC Supplier No. HY1342210MG
Zatebradine hydrochloride is a research-grade small molecule used to inhibit hyperpolarization-activated cyclic nucleotide-gated (HCN) channels in electrophysiology and pharmacology studies. It blocks HCN1-HCN4-mediated currents with low-micromolar potency and is supplied in solid and solution formats for in vitro experiments.
  • Potent HCN channel antagonist with low-micromolar IC50 values.
  • Blocks HCN1-HCN4-mediated slow inward current.
  • Available as solid material and as a 10 mM solution in DMSO for convenience.
  • High reported purity suitable for research applications.
  • Applicable to electrophysiology, cardiac research, and ion channel pharmacology studies.