Learn More
Medchemexpress LLC HY-13422 5mg Medchemexpress, Zatebradine (hydrochloride) CAS:91940-87-3 Purity:>98%
SDP
Catalog No. 502081361
Encompass_Preferred
Medchemexpress, HY-13422 5mg Zatebradine (hydrochloride) CAS:91940-87-3 Zatebradine (UL-FS-49 (free base)) is a potent inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels with an IC50 values 1.96 µM. Zatebradine blocks the slow inward current through human HCN1, HCN2, HCN3 and HCN4 channels, with IC50 values of 1.83 µM, 2.21 µM, 1.90 µM and 1.88 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1361 Supplier Medchemexpress LLC Supplier No. HY134225MG
Medchemexpress, HY-13422 5mg Zatebradine (hydrochloride) CAS:91940-87-3 Zatebradine (UL-FS-49 (free base)) is a potent inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels with an IC50 values 1.96 µM. Zatebradine blocks the slow inward current through human HCN1, HCN2, HCN3 and HCN4 channels, with IC50 values of 1.83 µM, 2.21 µM, 1.90 µM and 1.88 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.