Learn More
Electron Microscopy Sciences Z-Axis Electrically Conductive Tape 9703, 1/2" wide 36 YD
SDP
Catalog No. 501901657
Encompass_Preferred
3M Electrically Conductive Adhesive Transfer Tape 9703 is a pressure sensitive adhesive (PSA) transfer tape with anisotropic electrical conductivity. The PSA matrix is filled with conductive particles which allow interconnection between substrates through the adhesive thickness (the "Z-axis") but are spaced far enough apart for the product to be electrically insulating in the plane of the adhesive. The PSA tack properties and lack of any thermal curing make tape 9703 easy to use in all applications requiring a conductive tape.
Width: ½"
Catalog No. 50-190-1657 Supplier Electron Microscopy Sciences Supplier No. 7780980
3M Electrically Conductive Adhesive Transfer Tape 9703 is a pressure sensitive adhesive (PSA) transfer tape with anisotropic electrical conductivity. The PSA matrix is filled with conductive particles which allow interconnection between substrates through the adhesive thickness (the "Z-axis") but are spaced far enough apart for the product to be electrically insulating in the plane of the adhesive. The PSA tack properties and lack of any thermal curing make tape 9703 easy to use in all applications requiring a conductive tape.
Width: ½"