Learn More
Phenomenex Inc Phenomenex GC Capillary Column, ZB-1, 30 m, 0.53 mm
Supplier:  Phenomenex Inc 7HKG00139B
Encompass_Preferred
Zebron ZB-1 GC capillary column with a non-polar dimethylpolysiloxane stationary phase, 30 m length, 0.53 mm internal diameter, and 5.00 μm film thickness. It delivers low bleed (MS certified), low activity, and high efficiency for reliable separations, is rated to 370 °C, and is suited for petrochemical fingerprinting, simulated distillation, and routine quality-control analyses.
  • Excellent resolving power for critical pairs in complex petrochemical samples.
  • Low bleed suitable for mass spectrometry.
  • Low activity for consistent peak shape.
  • High efficiency for sharp, reproducible separations.
  • Rated to 370 °C maximum temperature for high-temperature applications.
  • Suitable for simulated distillation and routine quality control analyses.
Catalog No. NC3737955