Learn More
Zymo Research Corporation ZymoBIOMICS™ Microbial Community DNA Standard (200 ng)
SDP
Catalog No. 501251612
Encompass_Preferred
One of the major challenges in the emerging field of microbiomics is the bias and errors introduced in the complex workflows. Besides nucleic acid purification, bias also arises from sequencing library preparation and subsequent processes. The ZymoBIOMICS Microbial Community DNA Standard is designed to assess bias, errors and other artifacts after the step of nucleic acid purification. This DNA standard is created by pooling DNA extracted from pure cultures. It has an accurately defined composition, negligible impurities (0.01%), and contains genomes of a wide range of GC content (15% - 85%). This DNA standard is designed to have the same microbial composition with the cellular version, the ZymoBIOMICS Microbial Community Standard, so that they can be more powerful when working in tandem.
Catalog No. 50-125-1612 Supplier Zymo Research Corporation Supplier No. D6305
One of the major challenges in the emerging field of microbiomics is the bias and errors introduced in the complex workflows. Besides nucleic acid purification, bias also arises from sequencing library preparation and subsequent processes. The ZymoBIOMICS Microbial Community DNA Standard is designed to assess bias, errors and other artifacts after the step of nucleic acid purification. This DNA standard is created by pooling DNA extracted from pure cultures. It has an accurately defined composition, negligible impurities (0.01%), and contains genomes of a wide range of GC content (15% - 85%). This DNA standard is designed to have the same microbial composition with the cellular version, the ZymoBIOMICS Microbial Community Standard, so that they can be more powerful when working in tandem.