Learn More
Aviva Systems Biology Corp ZBED1 Antibody - C-terminal region (ARP50502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBED1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZBED1. Alternate names: ALTE, DREF, KIAA0785, TRAMP, hDREFApplicationWB. Species reactivity: Cow, Horse, Human, Rabbit, Yeast
Catalog No. 50-285-513