Learn More
Aviva Systems Biology Corp ZBP1 Antibody - middle region (ARP36711_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36711_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZBP1. Alternate names: DAI, DLM1, DLM-1, C20orf183ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53845