Learn More
Uline Zebra ZD621D Desktop Direct Thermal Barcode Printer,1 EA
Catalog No. NC2595000
Encompass
Prints super fast � up to 8" per second. Easy to load in one step. Efficient design prevents label jams. Increased memory for faster processing. Compatible with Windows-based software/systems. Not compatible with UPS WorldShip or FedEx Software. Supports EPL and ZPL printer languages. Use Bluetooth or Ethernet interface to connect to a network. Use with Desktop Direct Thermal Barcode Labels and Barcode Label Software. Buying Guide available.
Catalog No. NC2595000 Supplier Uline Supplier No. H9607
Prints super fast � up to 8" per second. Easy to load in one step. Efficient design prevents label jams. Increased memory for faster processing. Compatible with Windows-based software/systems. Not compatible with UPS WorldShip or FedEx Software. Supports EPL and ZPL printer languages. Use Bluetooth or Ethernet interface to connect to a network. Use with Desktop Direct Thermal Barcode Labels and Barcode Label Software. Buying Guide available.