Learn More
Aviva Systems Biology Corp ZBTB10 Antibody - middle region (ARP36027_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36027_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZBTB10. Alternate names: FLJ12752, RINZFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55976