Learn More
Aviva Systems Biology Corp ZBTB10 Antibody - N-terminal region (ARP37785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB10. Alternate names: FLJ12752, RINZFApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-56431