Learn More
Aviva Systems Biology Corp ZBTB11 Antibody - N-terminal region (ARP39083_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39083_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB11. Alternate names: FLJ13426, MGC133303, ZNF-U69274, ZNF913ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56906