Learn More
Aviva Systems Biology Corp ZBTB12 Antibody - N-terminal region (ARP40042_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40042_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB12. Alternate names: Bat9, C6orf46, D6S59E, G10, NG35ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-57275