Learn More
Aviva Systems Biology Corp ZBTB16 Antibody - C-terminal region (ARP38723_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38723_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZBTB16. Alternate names: PLZF, ZNF145ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56769