Learn More
Aviva Systems Biology Corp ZBTB20 Antibody - middle region (ARP31853_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31853_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZBTB20. Alternate names: HOF, DPZF, ODA-8S, ZNF288ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52839