Learn More
Aviva Systems Biology Corp ZBTB25 Antibody - middle region (ARP38890_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38890_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZBTB25. Alternate names: KUP, ZNF46, C14orf51ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-56836