Learn More
Aviva Systems Biology Corp ZBTB26 Antibody - N-terminal region (ARP39457_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39457_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB26. Alternate names: KIAA1572, ZNF481, biorefApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57038