Learn More
Aviva Systems Biology Corp ZBTB39 Antibody - middle region (ARP33688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZBTB39. Alternate names: KIAA0352, ZNF922ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-55117