Learn More
Aviva Systems Biology Corp Zbtb43 Antibody - C-terminal region (ARP39048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zbtb43. It was validated on Western Blot. . Alternate names: 1700010E06Rik, Zfp297b, mKIAA0414ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-776