Learn More
Aviva Systems Biology Corp ZBTB44 Antibody - middle region (ARP35875_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35875_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB44. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZBTB44. Alternate names: BTBD15, HSPC063, MGC26123, MGC57431, MGC60348, MGC88058, ZNF851ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55902