Learn More
Aviva Systems Biology Corp ZBTB6 Antibody - N-terminal region (ARP38836_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38836_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB6. Alternate names: ZID, ZNF482ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53115