Learn More
Aviva Systems Biology Corp ZBTB7A Antibody - middle region (ARP35899_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35899_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB7A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZBTB7A. Alternate names: LRF, FBI1, FBI-1, ZBTB7, ZNF857A, pokemonApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-55916