Learn More
Aviva Systems Biology Corp ZBTB7A Antibody - N-terminal region (ARP37084_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37084_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB7A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse ZBTB7A. Alternate names: Lrf, FBI-1, Zbtb7, Pokemon, AI452336, 9030619K07Rik, 9130006G12RikApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56321