Learn More
Aviva Systems Biology Corp ZBTB7A Antibody - N-terminal region (ARP39211_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39211_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB7A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB7A. Alternate names: DKFZp547O146, FBI-1, FBI1, LRF, MGC99631, ZBTB7, pokemon, ZNF857AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56943