Learn More
Aviva Systems Biology Corp Zbtb7a Antibody - C-terminal region (ARP39212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zbtb7a. It was validated on Western Blot. . Alternate names: 9030619K07Rik, 9130006G12Rik, AI452336, FBI-1, Lrf, Pokemon, Zbtb7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-284-786