Learn More
Aviva Systems Biology Corp ZBTB7C Antibody - N-terminal region (ARP32872_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32872_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB7C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB7C. Alternate names: APM1, APM-1, ZBTB36, ZNF857CApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54802