Learn More
Aviva Systems Biology Corp Zc3h10 Antibody - N-terminal region (ARP60671_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60671_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zc3h10. It was validated on Western Blot. . Alternate names: AI790326ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-122