Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci ZC3H12A (CT) Peptide
SDP

Catalog No. 16595549
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-549 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-549 Supplier ProSci Supplier No. 9547P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

ZC3H12A (CT) Peptide is used for blocking the activity of ZC3H12A (CT) Antibody.

ZC3H12A Antibody: ZC3H12A is an MCP1 (CCL2; MIM 158105)-induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis. ZC3H12A inhibits the Wnt/beta-catenin signaling pathway activity and modulates epithelial-mesenchymal transition during clear cell renal cell carcinoma progression by targeting miRNAs. ZC3H12A ribonuclease can bind and cleave AURKA mRNA in MYCN-amplified neuroblastoma cells. ZC3H12A is a novel link between diabetogenic conditions and impaired insulin secretory capacity. ZC3H12A overexpression in human neuroblastoma cell lines causes cell-cycle arrest by G1/S checkpoint block.

Specifications

Gene ID (Entrez) 80149
Species Human
Accession Number Q5D1E7.2
Common Name zinc finger CCCH-type containing 12 A
Concentration 200 μg/mL
Content And Storage Store SARS-CoV-2 (COVID-19) Spike Q493R G496S Q498R N501Y Y505H Peptide (Omicron) at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol ZC3H12A
Quantity 0.05 mg
Sequence 14 amino acids near the carboxyl terminus of human ZC3H12A.
Synonym ZC3H12A Antibody: Endoribonuclease ZC3H12A, Monocyte chemotactic protein-induced protein 1, MCP-induced protein 1, MCPIP-1, Zinc finger CCCH domain-containing protein 12 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.