Learn More
Aviva Systems Biology Corp ZC3H15 Antibody - middle region (ARP50635_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50635_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZC3H15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZC3H15. Alternate names: HT010, LEREPO4, MSTP012ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60393