Learn More
ABclonal Technology ZC3H7A Rabbit pAb
SDP
Supplier:  ABclonal Technology A13190500UL
Encompass_Preferred
Enables miRNA binding activity. Involved in production of miRNAs involved in gene silencing by miRNA. Predicted to be located in nucleus.
Catalog No. 50-001-16483