Learn More
Aviva Systems Biology Corp ZC3H7B Antibody - middle region (ARP31808_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZC3H7B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZC3H7B. Alternate names: DKFZp434K0920, FLJ13787, KIAA1031, RoXaNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54328