Learn More
Aviva Systems Biology Corp TUT4 Antibody - middle region (ARP58745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZCCHC11. It was validated on Western Blot. ZCCHC11,zinc finger, CCHC domain containing 11,TUT4; PAPD3; FLJ42878; DKFZp779C1943; ZCCHC11,terminal uridylyltransferase 4,terminal uridylyltransferase 4,TUTase 4,PAP associated domain containing 3,zinc finger CCHC domain-containing protein 11The immunogen is a synthetic peptide directed towards the middle region of human ZCCHC11. Alternate names: PAPD3, TUT4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-494