Learn More
Aviva Systems Biology Corp ZCCHC12 Antibody - N-terminal region (ARP53282_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53282_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZCCHC12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZCCHC12. Alternate names: FLJ16123, SIZN, SIZN1, PNMA7AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53708