Learn More
Aviva Systems Biology Corp ZCCHC3 Antibody - middle region (ARP50731_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50731_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZCCHC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZCCHC3. Alternate names: C20orf99, MGC104290ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60420