Learn More
Aviva Systems Biology Corp ZCCHC7 Antibody - C-terminal region (ARP60538_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60538_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZCCHC7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZCCHC7. Alternate names: AIR1, FLJ22611, HSPC086, RP11-397D12.1ApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-287-079