Learn More
Selleck Chemical LLC ZCL278 10mg 587841-73-4
SDP
Supplier:  Selleck Chemical LLC S729310MG
Encompass_Preferred
ZCL278 is a selective Cdc42 GTPase inhibitor with Kd of 11.4 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4605