Learn More
Medchemexpress LLC HY-13963 100mg , ZCL278 CAS:587841-73-4 Purity:98%
SDP
Catalog No. 501493429
Encompass_Preferred
Medchemexpress, HY-13963 100mg ZCL278 CAS:587841-73-4 Formula:C21H19BrClN5O4S2 Kd: 11.4 μM (Cdc42) Purity:98% ZCL278 is a selective Cdc42 modulator that directly binds to Cdc42 and inhibits its functions with K d of 11.4 μM for Cdc42-ZCL278 affinity in surface plasmon resonance (SPR) experiment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3429 Supplier Medchemexpress LLC Supplier No. HY13963100MG
Medchemexpress, HY-13963 100mg ZCL278 CAS:587841-73-4 Formula:C21H19BrClN5O4S2 Kd: 11.4 μM (Cdc42) Purity:98% ZCL278 is a selective Cdc42 modulator that directly binds to Cdc42 and inhibits its functions with K d of 11.4 μM for Cdc42-ZCL278 affinity in surface plasmon resonance (SPR) experiment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.