Learn More
Zymo Research Corporation Mix & Go! Competent Cells-XJb(DE3) Autolysis™ (10 x 100 μl) (Add 1 ml 500x Arabinose)
SDP
Catalog No. 50444646
Encompass_Preferred
Additionally, it does not require use of any potentially interferingcomponents such as detergents, commonly found in various lytic buffers.Highlights Simple and controlled autolysis of E. coli Strains can by lysed in minutes after harvesting Method compatible with most buffer systems Ideal for protein expression and purification, also applicable for extraction of nucleic acids Scale up to lyse more samples without increase in time or labor
Catalog No. 50-444-646 Supplier Zymo Research Corporation Supplier No. T3051
Additionally, it does not require use of any potentially interferingcomponents such as detergents, commonly found in various lytic buffers.Highlights Simple and controlled autolysis of E. coli Strains can by lysed in minutes after harvesting Method compatible with most buffer systems Ideal for protein expression and purification, also applicable for extraction of nucleic acids Scale up to lyse more samples without increase in time or labor