Learn More
Medchemexpress LLC ZD-0892 | 171964-73-1 | 95.0% | 499.52 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1925410MM/1ML
Encompass_Preferred
ZD-0892 is a selective and potent inhibitor of neutrophil elastase, with Ki values of 6.7 nM for human neutrophil elastase and 200 nM for porcine pancreatic elastase. It is intended for research use only and not for sale to patients. The product is a solid, white to off-white in appearance.
  • Selective and potent inhibitor of neutrophil elastase.
  • Reduces myocardial elastolytic activity and coronary microvascular perfusion injury in DBA/2 mice infected with the EMC virus.
  • Decreases the pathologic severity of myocardial lesions associated with murine viral myocarditis.
  • Capable of reversing advanced pulmonary vascular disease in rats.
Catalog No. 50-003-85436