Learn More
Medchemexpress LLC ZD-0892 | 171964-73-1 | 95.0% | 499.52 | 25 MG
SDP
Catalog No. 5000385939
Encompass_Preferred
ZD-0892 is a selective and potent inhibitor of neutrophil elastase. It exhibits high affinity for human neutrophil elastase and porcine pancreatic elastase, making it suitable for targeted research.
  • Selective and potent inhibitor of neutrophil elastase
  • Suitable for in vitro and in vivo studies
  • Available in solid form for various research needs
  • Backed by scientific publications for its applications
Catalog No. 50-003-85939 Supplier Medchemexpress LLC Supplier No. 25MGHY19254
ZD-0892 is a selective and potent inhibitor of neutrophil elastase. It exhibits high affinity for human neutrophil elastase and porcine pancreatic elastase, making it suitable for targeted research.
  • Selective and potent inhibitor of neutrophil elastase
  • Suitable for in vitro and in vivo studies
  • Available in solid form for various research needs
  • Backed by scientific publications for its applications