Learn More
Medchemexpress LLC HY-19254 5mg Medchemexpress, ZD-0892 CAS:171964-73-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY192545MG
Encompass_Preferred
Medchemexpress, HY-19254 5mg ZD-0892 CAS:171964-73-1 ZD-0892 is a selective and potent inhibitor of a neutrophil elastase with Kis of 6.7 and 200 nM for human neutrophil elastase and porcine pancreatic elastase, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2110